4-tert-Butyl-o-xylene ≥98.0%
Supplier: TCI America
Synonyms:
1,2-Dimethyl-4-tert-butylbenzene
, 1-tert-Butyl-3,4-dimethylbenzene
B1338-25G
TCB1338-025GEA
35.48
USD
TCB1338-025G
4-tert-Butyl-o-xylene ≥98.0%
4-tert-Butyl-o-xylene
CAS Number: 7397-06-0
MDL Number: MFCD00038277
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 211
Flash Point (°C): 82
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: 4-tert-Butyl-1,2-dimethylbenzene
Formula:
C₁₂H₁₈ MW: 162.27 g/mol Boiling Pt: 211 °C Density: 0.87 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00038277 CAS Number: 7397-06-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...