4-(Benzyloxy)benzaldehyde
Supplier: TCI America
B1325-25G
TCB1325-025GEA
53.8
USD
TCB1325-025G
4-(Benzyloxy)benzaldehyde
4-(Benzyloxy)benzaldehyde
CAS Number: 4397-53-9
MDL Number: MFCD00003387
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Form: Crystal
Color: White
Boiling point (°C): 219
Melting point (°C): 74
Formula:
C₁₄H₁₂O₂ MW: 212.25 g/mol Boiling Pt: 219 °C Melting Pt: 74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003387 CAS Number: 4397-53-9 EINECS: 224-527-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...