4-Bromo-2-fluoroaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B1314-25G
TCB1314-025GEA 73.08 USD
TCB1314-025G
4-Bromo-2-fluoroaniline ≥98.0% (by GC, titration analysis)
4-Bromo-2-fluoroaniline

CAS Number: 367-24-8
MDL Number: MFCD00010221
Molecular Formula: C6H5BrFN
Molecular Weight: 190.02
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 122
Melting point (°C): 40
Flash Point (°C): 104

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 122 °C
Melting Pt: 40 °C
Flash Pt: 104 °C
MDL Number: MFCD00010221
CAS Number: 367-24-8
EINECS: 206-685-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR