4-Bromo-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Bromo-2-methylaniline2-Amino-5-bromotoluene

B1281-25G
TCB1281-025GEA 43.65 USD
TCB1281-025G
4-Bromo-o-toluidine ≥98.0% (by GC, titration analysis)
4-Bromo-o-toluidine

CAS Number: 583-75-5
MDL Number: MFCD00007825
Molecular Formula: C7H8BrN
Molecular Weight: 186.05
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 240
Melting point (°C): 60
Flash Point (°C): 113
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Amino-5-bromotoluene, 4-Bromo-o-toluidine

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 240 °C
Melting Pt: 60 °C
Flash Pt: 113 °C
MDL Number: MFCD00007825
CAS Number: 583-75-5
EINECS: 209-519-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR