4,4'-Bis(diethylamino)benzophenone ≥92.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4,4-Bis(diethylamino) benzophenoneN,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone

B1275-25G B1275-500G
TCB1275-025GEA 32.48 USD
TCB1275-025G TCB1275-500G
4,4'-Bis(diethylamino)benzophenone ≥92.0% (by HPLC, titration analysis)
4,4'-Bis(diethylamino)benzophenone

CAS Number: 90-93-7
MDL Number: MFCD00009044
Molecular Formula: C21H28N2O
Molecular Weight: 324.47
Purity/Analysis Method: 92.0% (HPLC,T)
Form: Crystal
Flash Point (°C): 151
Lambda max.: 352 nm (THF)


Other Product Names / Synonyms: N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone

Formula: C₂₁H₂₈N₂O
MW: 324.47 g/mol
Boiling Pt: 420 °C (1013 hPa)
Melting Pt: 96.5 °C
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00009044
CAS Number: 90-93-7
EINECS: 202-025-4
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/31/08/28933108.jpg

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR