N-tert-Butylformamide ≥98.0%
Supplier: TCI America
Synonyms:
N-tert-butylformamide
, N-Formyl-tert-butylamine
B1273-25ML
TCB1273-025MLEA
91.35
USD
TCB1273-025ML
N-tert-Butylformamide ≥98.0%
N-tert-Butylformamide
CAS Number: 2425-74-3
MDL Number: MFCD00003277
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 202
Flash Point (°C): 82
Freezing point (°C): 13
Specific Gravity (20/20): 0.90
Other Product Names / Synonyms: N-Formyl-tert-butylamine
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 202 °C Density: 0.90 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00003277 CAS Number: 2425-74-3 EINECS: 219-369-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...