(R)-(+)-1,1'-Bi-2-naphthol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (R)-BINOL(R)-(+)-1,1'-Binaphthyl-2,2'-Diol(R)-[1,1'-Binaphthalene]-2,2'-diol

B1142-25G B1142-5G
TCB1142-005GEA 64 USD
TCB1142-005G TCB1142-025G
(R)-(+)-1,1'-Bi-2-naphthol ≥98.0% (by HPLC, titration analysis)
(R)-(+)-1,1'-Bi-2-naphthol

CAS Number: 18531-94-7
MDL Number: MFCD00004068
Molecular Formula: C20H14O2
Molecular Weight: 286.33
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 210
Specific rotation [a]20/D: 36 deg (C=1, THF)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Asymmetric Synthesis
Other Product Names / Synonyms: (R)-(+)-BINOL, (R)-(+)-2,2'-Dihydroxy-1,1'-binaphthyl

Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00004068
CAS Number: 18531-94-7
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/29/86/28932986.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR