Bis-trispropane ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
[for Buffer Material]
CAS Number: 64431-96-5
MDL Number: MFCD00004689
Molecular Formula: C11H26N2O6
Molecular Weight: 282.34
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 166
Product Categories: Bioscience, Buffer-related Reagents
Other Product Names / Synonyms: BIS-TRIS propane
Formula:
C₁₁H₂₆N₂O₆ MW: 282.34 g/mol Melting Pt: 166 °C |
MDL Number:
MFCD00004689 CAS Number: 64431-96-5 EINECS: 264-899-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...