4-Bromoveratrole ≥97.0%
Supplier: TCI America
Synonyms:
1-Bromo-3,4-dimethoxybenzene
, 4-Bromo-1,2-dimethoxybenzene
CAS Number: 2859-78-1
MDL Number: MFCD00008381
Molecular Formula: C8H9BrO2
Molecular Weight: 217.06
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 256
Flash Point (°C): 109
Specific Gravity (20/20): 1.51
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 4-Bromocatechol Dimethyl Ether, 4-Bromoveratrole
Formula:
C₈H₉BrO₂ MW: 217.06 g/mol Boiling Pt: 256 °C Density: 1.51 (20 °C) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008381 CAS Number: 2859-78-1 EINECS: 220-677-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...