α-Bromo-m-xylene ≥90.0%
Supplier: TCI America
Synonyms:
3-Methylbenzyl bromide
B1010-25G
TCB1010-25GEA
66.99
USD
TCB1010-25G
α-Bromo-m-xylene ≥90.0%
α-Bromo-m-xylene
CAS Number: 620-13-3
MDL Number: MFCD00000177
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 82
Specific Gravity (20/20): 1.37
Other Product Names / Synonyms: 3-Methylbenzyl Bromide
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 98 °C Melting Pt: 18 °C Density: 1.37 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00000177 CAS Number: 620-13-3 EINECS: 210-625-2 UN: 1701 ADR: 6.1,II Merck Index: 12,10223 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...