4-Bromo-m-xylene ≥97.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2,4-Dimethylbromobenzene
, 1-Bromo-2,4-dimethylbenzene
B0996-25G
B0996-250G
TCB0996-250GEA
770.44
USD
TCB0996-250G
TCB0996-025G
4-Bromo-m-xylene ≥97.0%
4-Bromo-m-xylene
CAS Number: 583-70-0
MDL Number: MFCD00000071
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 79
Specific Gravity (20/20): 1.36
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 208 °C Melting Pt: –1 °C Density: 1.36 (20 °C) Flash Pt: 79 °C |
MDL Number:
MFCD00000071 CAS Number: 583-70-0 EINECS: 209-517-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...