2-Bromo-1,4-dichlorobenzene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Bromo-2,5-dichlorobenzene

B0981-25G
TCB0981-25GEA 87.29 USD
TCB0981-25G
2-Bromo-1,4-dichlorobenzene ≥98.0%
2-Bromo-1,4-dichlorobenzene

CAS Number: 1435-50-3
MDL Number: MFCD00018505
Molecular Formula: C6H3BrCl2
Molecular Weight: 225.89
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Flash Point (°C): 113
Freezing point (°C): 31

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 120 °C
Melting Pt: 31…33 °C
Flash Pt: 113 °C
MDL Number: MFCD00018505
CAS Number: 1435-50-3
EINECS: 215-859-9
https://us.vwr.com/stibo/web/std.lang.all/28/50/28932850.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR