(1-Butyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Butyltriphenylphosphonium bromide
B0970-25G
B0970-250G
TCB0970-025GEA
34.43
USD
TCB0970-025G
TCB0970-250G
(1-Butyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
(1-Butyl)triphenylphosphonium bromide
CAS Number: 1779-51-7
MDL Number: MFCD00011855
Molecular Formula: C22H24BrP
Molecular Weight: 399.31
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 243
Formula:
H₃C(CH₂)₃P(C₆H₅)₃Br MW: 399.31 g/mol Melting Pt: 243 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011855 CAS Number: 1779-51-7 EINECS: 217-219-4 UN: 3464 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...