Benzylidenemalononitrile ≥98.0%

Supplier: TCI America

B0947-25G B0947-10G
TCB0947-25GEA 141.71 USD
TCB0947-25G TCB0947-010G
Benzylidenemalononitrile ≥98.0%
Benzylidenemalononitrile

CAS Number: 2700-22-3
MDL Number: MFCD00001855
Molecular Formula: C10H6N2
Molecular Weight: 154.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: Benzylidenemalononitrile

Formula: C₁₀H₆N₂
Melting Pt: 85 °C
MDL Number: MFCD00001855
CAS Number: 2700-22-3
EINECS: 220-283-6
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/28/26/28932826.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR