4-Butoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0896-25G
TCB0896-025GEA 60.9 USD
TCB0896-025G
4-Butoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Butoxybenzoic acid

CAS Number: 1498-96-0
MDL Number: MFCD00002546
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 162

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 162 °C
MDL Number: MFCD00002546
CAS Number: 1498-96-0
EINECS: 216-104-6
https://us.vwr.com/stibo/web/std.lang.all/27/84/28932784.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR