p-Benzoquinone ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Quinone 1,4-Benzoquinone

B0887-25G B0887-500G
TCB0887-500GEA 103.53 USD
TCB0887-500G TCB0887-25G
p-Benzoquinone ≥97.0% (by titrimetric analysis)
p-Benzoquinone

CAS Number: 106-51-4
MDL Number: MFCD00001591
Molecular Formula: C6H4O2
Molecular Weight: 108.10
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 113
Flash Point (°C): 78


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction), Materials Chemistry, Polymerization Inhibitors
Other Product Names / Synonyms: p-Quinone, p-Benzoquinone

Formula: C₆H₄O₂
MW: 108.1 g/mol
Boiling Pt: ∼174 °C (1013 hPa)
Melting Pt: 113 °C
Density: 1.318 g/cm³ (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00001591
CAS Number: 106-51-4
EINECS: 203-405-2
UN: 2587
ADR: 6.1,II
Merck Index: 12,08259
https://us.vwr.com/stibo/web/std.lang.all/27/78/28932778.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR