1-Bromo-4-fluorobenzene ≥98.0%
Supplier: TCI America
Synonyms:
4-Bromofluorobenzene
B0884-100G
B0884-25G
B0884-500G
TCB0884-25GEA
31.47
USD
TCB0884-25G
TCB0884-100G
TCB0884-500G
1-Bromo-4-fluorobenzene ≥98.0%
1-Bromo-4-fluorobenzene
CAS Number: 460-00-4
MDL Number: MFCD00000342
Molecular Formula: C6H4BrF
Molecular Weight: 175.00
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 56
Specific Gravity (20/20): 1.60
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Benzyne Precursors (Aryne Precursors), Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 1-Bromo-4-fluorobenzene
Formula:
C₆H₄BrF MW: 175 g/mol Boiling Pt: 153 °C Melting Pt: –16 °C Density: 1.60 (20 °C) Flash Pt: 56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000342 CAS Number: 460-00-4 EINECS: 207-300-2 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...