tert-Butylcyclohexane ≥99.0%
Supplier: TCI America
B0823-25ML
TCB0823-025MLEA
44.64
USD
TCB0823-025ML
tert-Butylcyclohexane ≥99.0%
tert-Butylcyclohexane
CAS Number: 3178-22-1
MDL Number: MFCD00001471
Molecular Formula: C10H20
Molecular Weight: 140.27
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 42
Specific Gravity (20/20): 0.81
Formula:
C₁₀H₂₀ MW: 140.27 g/mol Boiling Pt: 171 °C Melting Pt: –41 °C Density: 0.81 (20 °C) Flash Pt: 42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001471 CAS Number: 3178-22-1 EINECS: 221-652-4 UN: 3295 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...