Butylcyclohexane ≥99.0%

Supplier: TCI America

B0822-500ML B0822-25ML
TCB0822-025MLEA 45.71 USD
TCB0822-025ML TCB0822-500ML
Butylcyclohexane ≥99.0%
Butylcyclohexane

CAS Number: 1678-93-9
MDL Number: MFCD00001530
Molecular Formula: C10H20
Molecular Weight: 140.27
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Melting point (°C): -78
Flash Point (°C): 41
Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: 1-Cyclohexylbutane

Formula: C₁₀H₂₀
Boiling Pt: 180 °C
Melting Pt: –78 °C
Density: 0.80 (20 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00001530
CAS Number: 1678-93-9
EINECS: 216-837-1
UN: 3295
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/27/30/28932730.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR