2-tert-Butyl-9,10-anthraquinone ≥98.0%

Supplier: TCI America

Synonyms: 2-tert-Butylanthraquinone

B0816-500G B0816-25G
TCB0816-025GEA 36.73 USD
TCB0816-025G TCB0816-500G
2-tert-Butyl-9,10-anthraquinone ≥98.0%
2-tert-Butyl-9,10-anthraquinone

CAS Number: 84-47-9
MDL Number: MFCD00001232
Molecular Formula: C18H16O2
Molecular Weight: 264.32
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 104

Formula: C₁₈H₁₆O₂
Melting Pt: 104 °C
MDL Number: MFCD00001232
CAS Number: 84-47-9
EINECS: 201-531-2
https://us.vwr.com/stibo/web/std.lang.all/27/26/28932726.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR