TCIAB0750

B0750-25ML B0750-500ML
TCB0750-25MLEA 53.43 USD
TCB0750-25ML TCB0750-500ML
TCIAB0750
CAS Number: 2028-63-9
MDL Number: MFCD00004541
Molecular Formula: C4H6O
Molecular Weight: 70.09
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 44
Specific Gravity (20/20): 0.95

Other Product Names / Synonyms: 1-Butyn-3-ol, Ethynylmethylcarbinol
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR