sec-Butylbenzene ≥99.0%

Supplier: TCI America

Synonyms: 2-Phenylbutane

B0714-500ML B0714-25ML
TCB0714-500MLEA 177.48 USD
TCB0714-500ML TCB0714-25ML
sec-Butylbenzene ≥99.0%
sec-Butylbenzene

CAS Number: 135-98-8
MDL Number: MFCD00009329
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Melting point (°C): -83
Flash Point (°C): 52
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 2-Phenylbutane

Formula: C₆H₅CH(CH₃)CH₂CH₃
MW: 134.22 g/mol
Boiling Pt: 173 °C
Melting Pt: –83 °C
Density: 0.86 (20 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00009329
CAS Number: 135-98-8
EINECS: 205-227-0
UN: 2709
ADR: 3,III
Merck Index: 13,01549
https://us.vwr.com/stibo/web/std.lang.all/26/46/28932646.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR