Butylbenzene ≥99.0%

Supplier: TCI America

Synonyms: n-Butylbenzene

B0713-25ML B0713-100ML B0713-500ML
TCB0713-25MLEA 53.64 USD
TCB0713-25ML TCB0713-100ML TCB0713-500ML
Butylbenzene ≥99.0%
Butylbenzene

CAS Number: 104-51-8
MDL Number: MFCD00009463
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Melting point (°C): -88
Flash Point (°C): 59
Specific Gravity (20/20): 0.86

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 183 °C
Melting Pt: –88 °C
Density: 0.86 (20 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00009463
CAS Number: 104-51-8
EINECS: 203-209-7
UN: 2709
ADR: 3,III
Merck Index: 13,01548
https://us.vwr.com/stibo/web/std.lang.all/26/45/28932645.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR