3-Bromo-α,α,α-trifluorotoluene ≥99.0%
Supplier: TCI America
Synonyms:
1-Bromo-3-(trifluoromethyl)benzene
B0664-25G
B0664-250G
TCB0664-25GEA
17.86
USD
TCB0664-25G
TCB0664-250G
3-Bromo-α,α,α-trifluorotoluene ≥99.0%
3-Bromo-α,α,α-trifluorotoluene
CAS Number: 401-78-5
MDL Number: MFCD00000380
Molecular Formula: C7H4BrF3
Molecular Weight: 225.01
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 43
Specific Gravity (20/20): 1.63
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Bromo-alpha,alpha,alpha-trifluorotoluene
Formula:
C₇H₄BrF₃ MW: 225.01 g/mol Boiling Pt: 155 °C Melting Pt: 37.5…39 °C Density: 1.63 (20 °C) Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000380 CAS Number: 401-78-5 EINECS: 206-932-6 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...