Ethyl-3-bromopropionate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 3-bromopropanoate

B0647-100G B0647-25G B0647-500G
TCB0647-025GEA 68 USD
TCB0647-025G TCB0647-500G TCB0647-100G
Ethyl-3-bromopropionate ≥98.0%
Ethyl 3-bromopropionate

CAS Number: 539-74-2
MDL Number: MFCD00000251
Molecular Formula: C5H9BrO2
Molecular Weight: 181.03
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 79
Specific Gravity (20/20): 1.42


Other Product Names / Synonyms: 3-Bromopropionic Acid Ethyl Ester

Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 136 °C
Density: 1.42 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00000251
CAS Number: 539-74-2
EINECS: 208-724-0
https://us.vwr.com/stibo/web/std.lang.all/25/84/28932584.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR