(±)-2-Bromopropionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: DL-2-Bromopropanoic acid2-Bromopropanoic acidDL-2-Bromopropionic acid(±)-2-Bromopropanoic acid

B0644-500G B0644-25G
TCB0644-500GEA 74.06 USD
TCB0644-500G TCB0644-25G
(±)-2-Bromopropionic acid ≥98.0% (by GC, titration analysis)
(±)-2-Bromopropionic acid

CAS Number: 598-72-1
MDL Number: MFCD00004211
Molecular Formula: C3H5BrO2
Molecular Weight: 152.98
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 100
Specific Gravity (20/20): 1.71

Formula: H₃CBrCHCOOH
MW: 152.98 g/mol
Boiling Pt: 205 °C
Melting Pt: ∼25 °C
Density: 1.71 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00004211
CAS Number: 598-72-1
EINECS: 209-947-6
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/25/79/28932579.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR