2-Bromoanisole ≥98.0%
Supplier: TCI America
Synonyms:
1-Bromo-2-methoxybenzene
B0546-100G
B0546-25G
B0546-500G
TCB0546-025GEA
44.67
USD
TCB0546-025G
TCB0546-500G
TCB0546-100G
2-Bromoanisole ≥98.0%
2-Bromoanisole
CAS Number: 578-57-4
MDL Number: MFCD00000064
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 223
Flash Point (°C): 101
Specific Gravity (20/20): 1.52
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 1-Bromo-2-methoxybenzene
Formula:
C₇H₇BrO MW: 187.04 g/mol Boiling Pt: 223 °C Density: 1.52 (20 °C) Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000064 CAS Number: 578-57-4 EINECS: 209-425-8 UN: 3082 ADR: 9,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...