4,4'-Biphenol ≥99.0%
Supplier: TCI America
Synonyms:
Biphenyl-4,4'-diol
B0464-100G
B0464-500G
B0464-25G
TCB0464-100GEA
140.01
USD
TCB0464-100G
TCB0464-500G
TCB0464-025G
4,4'-Biphenol ≥99.0%
4,4'-Biphenol
CAS Number: 92-88-6
MDL Number: MFCD00002348
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 281
Product Categories: Materials Chemistry, Monomers, Biphenyls (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Biphenyl-4,4'-diol, 4,4'-Biphenol
Formula:
C₁₂H₁₀O₂ MW: 186.21 g/mol Boiling Pt: 355 °C (1013 hPa) Melting Pt: 281 °C Density: 1.25 g/cm³ (20 °C) |
MDL Number:
MFCD00002348 CAS Number: 92-88-6 EINECS: 202-200-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...