4-Benzylpyridine ≥98.0%

Supplier: TCI America
Warning

B0437-25ML
TCB0437-025MLEA 23.26 USD
TCB0437-025ML
4-Benzylpyridine ≥98.0%
4-Benzylpyridine

CAS Number: 2116-65-6
MDL Number: MFCD00006443
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 287
Melting point (°C): 11
Flash Point (°C): 115
Specific Gravity (20/20): 1.06

Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 287 °C
Melting Pt: 11 °C
Density: 1.06 (20 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00006443
CAS Number: 2116-65-6
EINECS: 218-319-0
https://us.vwr.com/stibo/web/std.lang.all/24/04/28932404.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR