2-Benzylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0436-25ML
TCB0436-25MLEA 58 USD
TCB0436-25ML
2-Benzylpyridine ≥98.0% (by GC, titration analysis)
2-Benzylpyridine

CAS Number: 101-82-6
MDL Number: MFCD00006352
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 277
Melting point (°C): 10
Flash Point (°C): 125
Specific Gravity (20/20): 1.06

Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 277 °C
Melting Pt: 10 °C
Density: 1.06 (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00006352
CAS Number: 101-82-6
EINECS: 202-979-1
https://us.vwr.com/stibo/web/std.lang.all/24/03/28932403.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR