Benzyl chloride ≥99.0% stabilized

Danger

Synonyms: α-Chlorotoluene (Chloromethyl)benzene Chlorophenylmethane

Stabilization: Stabilised with Stabilised

B0412-25G B0412-500G
TCB0412-25GEA 15.3 USD
TCB0412-25G TCB0412-500G
Benzyl chloride ≥99.0% stabilized
Benzyl chloride

(stabilized with epsilon-Caprolactam)
CAS Number: 100-44-7
MDL Number: MFCD00000889
Molecular Formula: C7H7Cl
Molecular Weight: 126.58
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 179
Flash Point (°C): 67
Specific Gravity (20/20): 1.10


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: alpha-Chlorotoluene

Formula: (C₆H₅)CH₂Cl
MW: 126.59 g/mol
Boiling Pt: 179 °C
Melting Pt: –41.2 °C
Density: 1.10 (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00000889
CAS Number: 100-44-7
EINECS: 202-853-6
UN: 1738
ADR: 6.1,II
Merck Index: 13,01131

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR