2-Bromoadamantane ≥97.0%

Supplier: TCI America

B0380-5G
TCB0380-005GEA 48 USD
TCB0380-005G
2-Bromoadamantane ≥97.0%
2-Bromoadamantane

CAS Number: 7314-85-4
MDL Number: MFCD00074745
Molecular Formula: C10H15Br
Molecular Weight: 215.13
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 140


Other Product Names / Synonyms: 2-Adamantyl Bromide

Formula: C₁₀H₁₅Br
MW: 215.13 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 140 °C
MDL Number: MFCD00074745
CAS Number: 7314-85-4
EINECS: 230-777-3
https://us.vwr.com/stibo/web/std.lang.all/23/67/28932367.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR