TCIAB0271

B0271-25ML
TCB0271-025MLEA 154.23 USD
TCB0271-025ML
TCIAB0271
CAS Number: 2696-84-6
MDL Number: MFCD00007924
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 104
Specific Gravity (20/20): 0.95
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR