(±)-2-Bromopentane ≥85.0% (contains ca. 12% 3-Bromopentane)
|
Danger
|
Synonyms:
(±)-2-Pentyl bromide
B0232-500ML
TCB0232-500MLEA
441.93
USD
TCB0232-500ML
(±)-2-Bromopentane ≥85.0% (contains ca. 12% 3-Bromopentane)
(±)-2-Bromopentane
CAS Number: 107-81-3
MDL Number: MFCD00000160
Molecular Formula: C5H11Br
Molecular Weight: 151.05
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 32
Specific Gravity (20/20): 1.21
Other Product Names / Synonyms: 2-Pentyl Bromide
Formula:
H₃CBrCH(CH₂)₂CH₃ MW: 151.05 g/mol Boiling Pt: 117 °C Melting Pt: –95.6 °C Density: 1.21 (20 °C) Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000160 CAS Number: 107-81-3 EINECS: 203-521-3 UN: 2343 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...