Benzoin ≥99.0%, zone refined: number of passes 40

Supplier: TCI America

Synonyms: 2-Hydroxy-2-phenylacetophenone

B0222-ZONE
TCB0222-ZONEEA 370 USD
TCB0222-ZONE
Benzoin ≥99.0%, zone refined: number of passes 40
Benzoin

CAS Number: 119-53-9
MDL Number: MFCD00004496
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 344
Melting point (°C): 135


Product Categories: Materials Chemistry, Polymerization Initiators, Highly Purified Reagents
Other Product Names / Synonyms: 2-Hydroxy-2-phenylacetophenone

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 344 °C
Melting Pt: 135 °C
MDL Number: MFCD00004496
CAS Number: 119-53-9
EINECS: 204-331-3
https://us.vwr.com/stibo/web/std.lang.all/23/03/28932303.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR