syn-Benzaldehyde oxime ≥90.0%
Supplier: TCI America
Synonyms:
(E)-Benzaldehyde oxime
, alpha-Benzaldoxime
B0011-100G
B0011-500G
B0011-25G
TCB0011-100GEA
236.42
USD
TCB0011-100G
TCB0011-025G
TCB0011-500G
syn-Benzaldehyde oxime ≥90.0%
syn-Benzaldehyde oxime
CAS Number: 622-31-1
MDL Number: MFCD00002119
Molecular Formula: C7H7NO
Molecular Weight: 121.14
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 123
Flash Point (°C): 108
Freezing point (°C): 27
Formula:
C₇H₇NO MW: 121.14 g/mol Boiling Pt: 123 °C Flash Pt: 108 °C |
MDL Number:
MFCD00002119 CAS Number: 622-31-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...