3-Amino-4-methoxybenzanilide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1954-25G
TCA1954-25GEA 38.51 USD
TCA1954-25G
3-Amino-4-methoxybenzanilide ≥98.0% (by titrimetric analysis)
3-Amino-4-methoxybenzanilide

CAS Number: 120-35-4
MDL Number: MFCD00017166
Molecular Formula: C14H14N2O2
Molecular Weight: 242.28
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 156


Other Product Names / Synonyms: 3-Amino-p-anisanilide

Formula: C₁₄H₁₄N₂O₂
MW: 242.28 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00017166
CAS Number: 120-35-4
EINECS: 204-388-4
https://us.vwr.com/stibo/web/std.lang.all/00/76/28930076.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR