2-(Allyloxy)benzaldehyde ≥96.0%

Supplier: TCI America

A1515-5G A1515-25G
TCA1515-005GEA 80 USD
TCA1515-005G TCA1515-25G
2-(Allyloxy)benzaldehyde ≥96.0%
2-(Allyloxy)benzaldehyde

CAS Number: 28752-82-1
MDL Number: MFCD00014130
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.09


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Salicylaldehyde Allyl Ether

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Density: 1.09 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00014130
CAS Number: 28752-82-1
EINECS: 249-198-2
https://us.vwr.com/stibo/web/std.lang.all/95/79/28929579.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR