Apigenin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4',5,7-Trihydroxyflavone 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one

A1514-100MG
TCA1514-100MGEA 168.53 USD
TCA1514-100MG
Apigenin ≥98.0% (by HPLC)
Apigenin

CAS Number: 520-36-5
MDL Number: MFCD00006831
Molecular Formula: C15H10O5
Molecular Weight: 270.24
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 4',5,7-Trihydroxyflavone

Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
MDL Number: MFCD00006831
CAS Number: 520-36-5
EINECS: 208-292-3
https://us.vwr.com/stibo/web/std.lang.all/95/77/28929577.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR