1-Acetylpiperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1399-5G A1399-25G
TCA1399-005GEA 16.24 USD
TCA1399-005G TCA1399-025G
1-Acetylpiperazine ≥98.0% (by GC, titration analysis)
1-Acetylpiperazine

CAS Number: 13889-98-0
MDL Number: MFCD00058676
Molecular Formula: C6H12N2O
Molecular Weight: 128.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 31

Formula: C₆H₁₂N₂O
Flash Pt: 110 °C
MDL Number: MFCD00058676
CAS Number: 13889-98-0
EINECS: 237-659-0
https://us.vwr.com/stibo/web/std.lang.all/93/44/28929344.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR