2-Acetyl-7-methoxybenzofuran ≥98.0%

Supplier: TCI America

Synonyms: 1-(7-Methoxy-1-benzofuran-2-yl)-1-ethanone

A1317-10G
TCA1317-010GEA 132.78 USD
TCA1317-010G
2-Acetyl-7-methoxybenzofuran ≥98.0%
2-Acetyl-7-methoxybenzofuran

CAS Number: 43071-52-9
MDL Number: MFCD00016911
Molecular Formula: C11H10O3
Molecular Weight: 190.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 93


Other Product Names / Synonyms: 2-Acetyl-7-methoxycoumarone

Formula: C₁₁H₁₀O₃
Boiling Pt: 180 °C
Melting Pt: 93 °C
MDL Number: MFCD00016911
CAS Number: 43071-52-9
https://us.vwr.com/stibo/web/std.lang.all/92/20/28929220.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR