(±)-1-Aminoindan ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,3-Dihydro-1H-inden-1-amine(±)-1-Indanylamine

A1309-10G
TCA1309-010GEA 170.52 USD
TCA1309-010G
(±)-1-Aminoindan ≥98.0% (by GC, titration analysis)
(±)-1-Aminoindan

CAS Number: 34698-41-4
MDL Number: MFCD00003799
Molecular Formula: C9H11N
Molecular Weight: 133.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 94
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 1-Indanamine

Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 220 °C
Melting Pt: 1.5 °C
Density: 1.04 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003799
CAS Number: 34698-41-4
EINECS: 252-158-7
https://us.vwr.com/stibo/web/std.lang.all/92/09/28929209.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR