Acetonyltriphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(2-Oxopropyl)triphenylphosphonium chloride
A1305-25G
TCA1305-025GEA
37.56
USD
TCA1305-025G
Acetonyltriphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Acetonyltriphenylphosphonium chloride
CAS Number: 1235-21-8
MDL Number: MFCD00011813
Molecular Formula: C21H20ClOP
Molecular Weight: 354.81
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 249
Formula:
C₂₁H₂₀ClOP MW: 354.82 g/mol Melting Pt: 249 °C |
MDL Number:
MFCD00011813 CAS Number: 1235-21-8 EINECS: 214-974-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...