3-Amino-5-methyl-1H-pyrazole ≥96.0%

Supplier: TCI America

Synonyms: 3-Amino-5-methylpyrazole5-Methyl-1H-pyrazol-3-amine

A1133-25G
TCA1133-025GEA 123.83 USD
TCA1133-025G
3-Amino-5-methyl-1H-pyrazole ≥96.0%
3-Amino-5-methyl-1H-pyrazole

CAS Number: 31230-17-8
MDL Number: MFCD00075180
Molecular Formula: C4H7N3
Molecular Weight: 97.12
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 41
Storage Temperature: 0-10°C

Formula: C₄H₇N₃
MW: 97.12 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00075180
CAS Number: 31230-17-8
https://us.vwr.com/stibo/web/std.lang.all/89/67/28928967.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR