1-Acenaphthenol ≥97.0%
Supplier: TCI America
Synonyms:
1,2-Dihydroacenaphthylen-1-ol
A1057-5G
TCA1057-5GEA
435.31
USD
TCA1057-5G
1-Acenaphthenol ≥97.0%
1-Acenaphthenol
CAS Number: 6306-07-6
MDL Number: MFCD00003808
Molecular Formula: C12H10O
Molecular Weight: 170.21
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 146
Other Product Names / Synonyms: 1-Hydroxyacenaphthene
Formula:
C₁₂H₁₀O MW: 170.21 g/mol Melting Pt: 146 °C |
MDL Number:
MFCD00003808 CAS Number: 6306-07-6 EINECS: 228-618-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...