5-Amino-2-nitrobenzoic acid ≥99.0% (by HPLC), purified
Supplier: TCI America
A1048-100MG
TCA1048-100MGEA
105
USD
TCA1048-100MG
5-Amino-2-nitrobenzoic acid ≥99.0% (by HPLC), purified
5-Amino-2-nitrobenzoic acid
[for gamma-GT]
CAS Number: 13280-60-9
MDL Number: MFCD00035748
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method:
Form: Crystal
Formula:
C₇H₆N₂O₄ MW: 182.14 g/mol Boiling Pt: 481 °C (1013 hPa) Melting Pt: 234 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035748 CAS Number: 13280-60-9 EINECS: 236-283-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...