Allyltriphenylphosphonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1007-25G
TCA1007-025GEA 127.89 USD
TCA1007-025G
Allyltriphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Allyltriphenylphosphonium bromide

CAS Number: 1560-54-9
MDL Number: MFCD00011808
Molecular Formula: C21H20BrP
Molecular Weight: 383.27
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 225

Formula: C₂₁H₂₀BrP
Melting Pt: 225 °C
MDL Number: MFCD00011808
CAS Number: 1560-54-9
EINECS: 216-332-6
https://us.vwr.com/stibo/web/std.lang.all/87/56/28928756.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR