9-Anthrealdehyde ≥99.0%

Supplier: TCI America

Synonyms: 9-Anthracenecarboxaldehyde 9-Anthracenecarbaldehyde

A0779-25G A0779-100G
TCA0779-100GEA 221.27 USD
TCA0779-100G TCA0779-025G
9-Anthrealdehyde ≥99.0%
9-Anthrealdehyde

CAS Number: 642-31-9
MDL Number: MFCD00001254
Molecular Formula: C15H10O
Molecular Weight: 206.24
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 107


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 9-Anthraldehyde

Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 369 °C (1013 hPa)
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001254
CAS Number: 642-31-9
EINECS: 211-383-0
https://us.vwr.com/stibo/web/std.lang.all/85/19/28928519.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR