4-Pentylbenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A0741-250G A0741-5G A0741-25G
TCA0741-250GEA 352 USD
TCA0741-250G TCA0741-025G TCA0741-005G
4-Pentylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Pentylbenzoic acid

CAS Number: 26311-45-5
MDL Number: MFCD00002572
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 89


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Pentylbenzoic Acid

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 260.7 °C (1013 hPa)
Melting Pt: 89 °C
Density: ∼0.992 g/cm³ (20 °C)
MDL Number: MFCD00002572
CAS Number: 26311-45-5
EINECS: 247-607-9
https://us.vwr.com/stibo/web/std.lang.all/84/30/28928430.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR