Allyltrimethylsilane ≥98.0%

Supplier: TCI America

A0729-250ML A0729-25ML
TCA0729-25MLEA 60.03 USD
TCA0729-25ML TCA0729-250ML
Allyltrimethylsilane ≥98.0%
Allyltrimethylsilane

CAS Number: 762-72-1
MDL Number: MFCD00008635
Molecular Formula: C6H14Si
Molecular Weight: 114.26
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Flash Point (°C): 7
Specific Gravity (20/20): 0.72

Formula: H₂CCHCH₂Si(CH₃)₃
MW: 114.26 g/mol
Boiling Pt: 83 °C
Density: 0.72 (20 °C)
Flash Pt: 7 °C
MDL Number: MFCD00008635
CAS Number: 762-72-1
EINECS: 212-104-5
https://us.vwr.com/stibo/web/std.lang.all/83/91/28928391.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR